Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diisopropyl Phosphite 98.0+%, TCI America™
SDP

Catalog No. p-7134705 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 1809-20-7
Molecular Formula C6H14O3P-
Molecular Weight (g/mol) 165.149
MDL Number MFCD00117905
InChI Key IWEDBEZKWHRUJX-UHFFFAOYSA-N
Synonym diisopropylphosphite, acmc-1bsaz, phosphorous acid diisopropyl anion, phosphorous acid, bis 1-methylethyl ester
PubChem CID 21884976
IUPAC Name dipropan-2-yl phosphite
SMILES CC(C)OP([O-])OC(C)C

Specifications

Color Colorless
Boiling Point 79°C
Quantity 25 g
Formula Weight 166.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diisopropyl Phosphite
EINECSNumber (2)-2001&(2)-1961
RTECSNumber SZ7660000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.