Learn More
Medchemexpress LLC HY-100957 5mg Medchemexpress, Dilazep (dihydrochloride) CAS:20153-98-4 Purity:>98%
Medchemexpress, HY-100957 5mg Dilazep (diHCl) CAS:20153-98-4 Dilazep dihydrochloride is an inhibitor of adenosine uptake. Dilazep dihydrochloride has cerebral and coronary vasodilating action through enhancement of effect of adenosine. Dilazep dihydrochloride also inhibits the ischemic damage, platelet aggregation, and membrane transport of nucleosides. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.