Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dilithium Phthalocyanine 93.0+%, TCI America™
SDP

Catalog No. P10491G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:

Chemical Identifiers

CAS

25510-41-2

Molecular Formula

C32H18Li2N8

Molecular Weight (g/mol)

528.432

MDL Number

MFCD00059094

InChI Key

VEGOPYLKEARSAH-UHFFFAOYSA-N

Synonym

dilithium phthalocyanine

PubChem CID

66736323

SMILES

[Li].[Li].C1=CC=C2C(=C1)C3=NC4=C5C=CC=CC5=C(N4)N=C6C7=CC=CC=C7C(=N6)N=C8C9=CC=CC=C9C(=N8)N=C2N3

Specifications

Quantity

1 g

Formula Weight

526.42

Percent Purity

≥93.0% (N)

Physical Form

Crystalline Powder

Chemical Name or Material

Dilithium Phthalocyanine

TSCA

Yes

Recommended Storage

Refrigerator

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.