Learn More
Thermo Scientific™ Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories
Accurately dispense media and achieve powerful, quiet, reliable homogenization with maximum productivity in your sample preparation.
Supplier: Thermo Scientific™ DB4005A
Description
Achieve maximum productivity with the Thermo Scientific™ Diluflux™ Automated Gravimetric Dilutor and Homogenizer Laboratory Blender Accessories. Expand the flexibility and maintain long-term functionality of the two platforms with accessories that enable the use of different media formats (bottles/bags), enhance the user interface and support proper function of the system.
The easy-to-use Diluflux Gravimetric Dilutor allows the simultaneous use of up to two diluents by having dual dispensing heads that are fed by dual peristaltic pumps mounted on the unit. The reliable and powerful Homogenizer Laboratory Blender enables rapid and efficient sample homogenization with a small footprint and quiet operation. The accessories listed here enable:- Alternative receptacles (bottles instead of bags)
- Alternative media sources (broth bags/bottles)
- Additional/replacement bag stands for ease of preparation
- Bag storage for incubation
- Sample labeling
- Maintenance with replacement parts
Not all products are available for sale in all territories. Please inquire.
Specifications
5 L | |
1 | |
Bottle |
Diluflux Diluter, Homogenizer, Bottle | |
Diluflux™ Diluter and Homogenizer | |
5L bottle (autoclavable) |
For research use only.