Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimenhydrinate, MP Biomedicals™

Catalog No. p-4405478 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10 g
500 g

Chemical Identifiers

CAS 523-87-5
Molecular Formula C24H28ClN5O3
Molecular Weight (g/mol) 469.97
InChI Key NFLLKCVHYJRNRH-UHFFFAOYSA-N
Synonym dimenhydrinate, dramamine, vomex a, chloranautine, diphenhydrinate, anautine, travelin, amosyt, aviomarin, gravol
PubChem CID 10660
IUPAC Name 2-benzhydryloxy-N,N-dimethylethanamine;8-chloro-1,3-dimethyl-7H-purine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)NC(=N2)Cl.CN(C)CCOC(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Color White
Quantity 10 g
Formula Weight 470
Physical Form Powder
Chemical Name or Material Dimenhydrinate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.