Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethoxymethylphenylsilane 98.0+%, TCI America™
SDP

Catalog No. p-7127227 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
100 mL

Chemical Identifiers

CAS 3027-21-2
Molecular Formula C9H14O2Si
Molecular Weight (g/mol) 182.29
MDL Number MFCD00008343
InChI Key CVQVSVBUMVSJES-UHFFFAOYSA-N
Synonym dimethoxymethylphenylsilane, dimethoxy methyl phenyl silane, methylphenyldimethoxysilane, phenylmethyldimethoxysilane, silane, dimethoxymethylphenyl, dimethoxyphenylmethylsilane, fenyl-dimethoxy-methylsilan, dimethoxy methyl phenylsilane, fenyl-dimethoxy-methylsilan czech, methylphenyldimethoxy silane
PubChem CID 18193
IUPAC Name dimethoxy(methyl)phenylsilane
SMILES CO[Si](C)(OC)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 199°C
Quantity 25 mL
Formula Weight 182.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethoxymethylphenylsilane
RTECSNumber VV3645000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.