Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethoxymethylvinylsilane 94.0+%, TCI America™
SDP

Catalog No. p-7127707 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 16753-62-1
Molecular Formula C5H12O2Si
Molecular Weight (g/mol) 132.234
MDL Number MFCD00129170
InChI Key ZLNAFSPCNATQPQ-UHFFFAOYSA-N
Synonym vinylmethyldimethoxysilane, dimethoxymethylvinylsilane, dimethoxy methyl vinyl silane, silane, ethenyldimethoxymethyl, ethenyldimethoxymethylsilane, methyldimethoxyvinylsilane, vinyl methyldimethoxysilane, dimethoxy methyl vinylsilane #, methylvinyldimethoxysilane
PubChem CID 85589
IUPAC Name ethenyl-dimethoxy-methylsilane
SMILES CO[Si](C)(C=C)OC

Specifications

Color Colorless
Boiling Point 104°C
Quantity 25 g
UN Number 1993
Formula Weight 132.23
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethoxymethylvinylsilane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.