Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Azobenzene-4,4′-dicarboxylate 95.0+%, TCI America™
SDP

Catalog No. p-7117530 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 5320-91-2
Molecular Formula C16H14N2O4
Molecular Weight (g/mol) 298.298
MDL Number MFCD00496622
InChI Key PSNNNVCIKCUWKM-UHFFFAOYSA-N
PubChem CID 585660
IUPAC Name methyl 4-[(4-methoxycarbonylphenyl)diazenyl]benzoate
SMILES COC(=O)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)OC

Specifications

Melting Point 240°C
Quantity 1 g
Synonym Azobenzene-4,4′C-dicarboxylic Acid Dimethyl Ester, 4,4′C-Azodibenzoic Acid Dimethyl Ester, Dimethyl 4,4′C-Azodibenzoate
Formula Weight 298.30
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Azobenzene-4,4′-dicarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.