Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl carbonate, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019282 Shop All MilliporeSigma Supelco Products
Click to view available options
:
100 mg

Dimethyl carbonate is a unique molecule with versatile chemical reactivity. It is an environmentally benign building block and also a safe substitute for dimethyl sulfate or methyl halides.

Environmental

Specifications

Quantity 100 mg
CAS 616-38-6
Type Analytical Standard
Linear Formula (CH3 O)2 CO
MDL Number MFCD00008420
Synonym Carbonic acid dimethyl ester
Formula Weight 90.08
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Vapor Pressure 18 mmHg (21.1°C)
Vapor Density 3.1 (vs. air)
Density 1.069 g/mL (at 25°C)
Melting Point 2°C to 4°C
Boiling Point 90°C
Flash Point 16°C
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.