Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, BioSyn™, for biosynthesis and applications requiring low water content, >99.5%, Burdick & Jackson, Honeywell

Catalog No. 60048434 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
4x4 L
  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity

Specifications

Synonym DMSO, Methyl Sulfoxide
Grade Low Water
Density 1.0954 g/cm3
Packaging Glass Bottle
Boiling Point 189°C
Quantity 4x4 L
Recommended Storage Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.