Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dimethylformamide, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500mL
4L
Packaging:
HDPE Pressure Bottom Poly Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
Specifications
Specifications
Formula Weight | 73.09 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | HDPE Pressure Bottom Poly Bottle |
Specific Gravity | 1L = 0.95kg |
Flash Point | 58.00°C |
Quantity | 500mL |
Chemical Name or Material | Dimethylformamide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction