Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylamine Sulfate 98%, Thermo Scientific™

Catalog No. AC408871000
Click to view available options
Quantity:

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS

587-84-8

Molecular Formula

C12H13NO4S

Molecular Weight (g/mol)

267.30

MDL Number

MFCD00042016

InChI Key

IPZMDJYHJNHGML-UHFFFAOYSA-N

Synonym

diphenylamine sulfate, benzenamine, n-phenyl-, sulfate 1:1, usaf ek-743, n-phenylaniline; sulfuric acid, diphenylamine, hydrogen sulfate, diphenylammonium hydrogen sulphate, diphenylamine; sulfuric acid, diphenylaminesulfate, bis n-phenylanilinium sulphate, diphenylaminsulfat

PubChem CID

11486

IUPAC Name

N-phenylaniline; sulfuric acid

SMILES

OS(O)(=O)=O.N(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point

144°C to 148°C

Color

White

Infrared Spectrum

Authentic

Assay Percent Range

98%

Linear Formula

(C6H5)2NH·H2SO4

Quantity

100 g

Merck Index

15, 3347

Formula Weight

267.3

Percent Purity

≥97.5%

Physical Form

Powder

Chemical Name or Material

Diphenylamine sulfate, 98%

Hazard Statement

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Precautionary Statement

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.