Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylthiocarbazide 98.0+%, TCI America™
SDP

Catalog No. D09175G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 622-03-7
Molecular Formula C13H14N4S
Molecular Weight (g/mol) 258.34
MDL Number MFCD00046042
InChI Key BNSNUHPJRKTRNT-UHFFFAOYSA-N
Synonym 1,5-Diphenyl-3-thiocarbohydrazide
PubChem CID 3032337
IUPAC Name 1,3-bis(phenylamino)thiourea
SMILES S=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1

Specifications

Melting Point 155°C
Quantity 5 g
Formula Weight 258.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Diphenylthiocarbazide
RTECSNumber FF2800000
TSCA Yes
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.