Learn More
AdipoGen Diquat metabolite dipyridone
SDP
Supplier:  AdipoGen CDXD1098M010
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 35022-72-1. Formula C12H10N2O2. MW BD9837. Diquat metabolite dipyridone is a common degradation product of diquat. Diquat is a nonselective, defoliant, preharvest, and non-residual bipyridyl herbicide and crop desiccant, normally used as the dibromide salt, to control broad-leaved weeds and grasses. Its activity is based on the liberation of the superoxide anion radical and, subsequently, hydrogen peroxide, leading to tissue destruction by oxidative stress. This compound can be used as a reference compound for analytical studies.
Catalog No. 50-256-1014