Learn More
AdipoGen DiSC33
SDP
Supplier:  AdipoGen CDXD0007M250
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 53336-12-2. Formula C23H25IN2S2. MW 520.49. Synthetic. Carbocyanine dyes, particularly thiacyanines such as DiSC33 can inhibit respiration and may therefore be relatively cytotoxic. 3,3-dipropyl-thiadicarbocyanine iodide is sensitive to membrane potential, fluorescence response to depolarization depends on the staining concentration and detection method. Selected applications are -Calcium channels and other ion transport systems -Mitochondrial activity -Neurons and brain tissue -Membrane potential in intact yeast cells
Catalog No. 50-256-1327