Learn More
KYFORA BIO LLC DSPC 1 GR
Supplier:  KYFORA BIO LLC 264371
Encompass_Preferred
DSPC (Distearoylphosphatidylcholine) is a zwitterionic phospholipid that is used in the preparation of liposomes for transfection and drug delivery applications. DSPC functions to enhance encapsulation efficiency and liposome stability which enables greater effectiveness in bioprocessing. Liposomes containing DSPC have been found to have less cytotoxicity and therefore greater cell viability when compared to other phospholipid-based systems. DSPC has been receiving increasing attention as a valuable component in forming lipid nanoparticles (LNPs) in the growing field of mRNA vaccines. LNPs facilitate the safe and effective delivery of mRNA to target cells, thus enabling therapeutic action. Key application(s): transfection, drug delivery, cellular uptake, LNP, liposomes
Catalog No. 50-255-9783
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.