Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disulfiram, Tocris Bioscience™
GSA_VA

Catalog No. 380750 Shop All R&D Systems Products
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Reversibly stimulates SERCA Ca2+-ATPase; displays a range of other activities

Inhibitor of aldehyde dehydrogenase that displays antialcoholism activity. Shown to reversibly stimulate Ca2+-ATPase activity and inhibit V-ATPase (EC50 = 26 μM). Also inhibits expression of MMP-2 and MMP-9 and displays anti-invasive activity. Pyroptosis inhibitor; binds gasdermin D (GSDMD) and inhibits pore formation. Also inhibits SARS-CoV-2 main protease (Mpro; IC50 = 9.35 μM) and inhibits viral replication in Vero cells in vitro.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 97-77-8
Molecular Formula C10H20N2S4
Molecular Weight (g/mol) 296.524
InChI Key AUZONCFQVSMFAP-UHFFFAOYSA-N
Synonym disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd
PubChem CID 3117
ChEBI CHEBI:4659
IUPAC Name diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)SSC(=S)N(CC)CC

Specifications

Quantity 50 mg
Formula Weight 296.54
Percent Purity >98%
Chemical Name or Material Disulfiram
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.