Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-α-Bromophenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog No. AC107271000
Click to view available options
Quantity:
Packaging:

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS

4870-65-9

Molecular Formula

C8H7BrO2

Molecular Weight (g/mol)

215.04

InChI Key

WAKFRZBXTKUFIW-UHFFFAOYSA-N

Synonym

alpha-bromophenylacetic acid, bromo phenyl acetic acid, .alpha.-bromophenylacetic acid, bromophenylacetic acid, bromo-phenyl-acetic acid, bromo-phenylacetic acid, dl-.alpha.-bromophenylacetic acid, benzeneacetic acid, .alpha.-bromo, phenylbromoacetic acid, bromo phenyl aceticacid

PubChem CID

97919

IUPAC Name

2-bromo-2-phenylacetic acid

SMILES

C1=CC=C(C=C1)C(C(=O)O)Br

Specifications

Melting Point

82.0°C to 83.0°C

Assay Percent Range

96% min. (HPLC)

Packaging

Glass bottle

Linear Formula

C6H5CH(Br)CO2H

Quantity

100 g

Beilstein

09,451

Solubility Information

Solubility in water: insoluble

Formula Weight

215.04

Percent Purity

97%

Chemical Name or Material

DL-α-Bromophenylacetic acid

Hazard Category

GHS Signal Word: Danger

Hazard Statement

GHS H Statement
Causes severe skin burns and eye damage.

Precautionary Statement

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously wit

EINECSNumber

225-477-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.