Learn More
DL-α-Bromophenylacetic acid, 97%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 82.0°C to 83.0°C |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass bottle |
Linear Formula | C6H5CH(Br)CO2H |
Quantity | 5 g |
Beilstein | 09,451 |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 215.04 |
Percent Purity | 97% |
Chemical Name or Material | DL-α-Bromophenylacetic acid |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Gently wash with plenty of soap and water. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously wit |
EINECSNumber | 225-477-4 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.