Learn More
Medchemexpress LLC HY-112758 10mg Medchemexpress, DLin-KC2-DMA CAS:1190197-97-7 Purity:>98%
Supplier: Medchemexpress LLC HY11275810MG
Medchemexpress, HY-112758 10mg DLin-KC2-DMA CAS:1190197-97-7 DLin-KC2-DMA is a cationic/ionizable lipid for siRNA delivery. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.