Learn More
KYFORA BIO LLC DLin-MC3-DMA 10 MG
Supplier:  KYFORA BIO LLC 1449610
Encompass_Preferred
DLin-MC3-DMA is an ionizable cationic lipids used in the formulation of lipid nanoparticles (LNPs) encapsulating siRNA, mRNA, plasmid DNA, and other nucleic acids for delivery in vitro or in vivo. DLin-MC3-DMA, when used in combination with other lipids, has been shown to be an especially potent vehicle for siRNA delivery.Key application(s): siRNA delivery, plasmid DNA delivery, mRNA delivery
Catalog No. 50-255-9799
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.