Learn More
Tocris Bioscience™ DMNPE-4 AM-caged-calcium
Shop All R&D Systems ProductsSpecifications
Chemical Name or Material
Bis(acetoxymethyl) 3,12-bis(2-(acetoxymethoxy)-2-oxoethyl)-5-(4,5-dimethoxy-2-nitrophenyl)-6,9-dioxa-3,12-diazatetradecane-1,14-dioate
Quantity
5 mg
Molecular Formula
C34H47N3O22
Formula Weight
849.74
Purity
95%
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.