Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DMNPE-caged ATP diammonium salt, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
Description
DMNPE-caged ATP; quantum yield is 0.07. Brain penetrant.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 750.44 |
Chemical Name or Material | DMNPE-caged ATP diammonium salt |
Safety and Handling
Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction