Learn More
Biochain Institute Inc Express Cloning Checker Kit I: Yellow Solution, 1 ml/PK
Supplier: Biochain Institute Inc K50112002
BioChain's Express Cloning Checker Kits provide highly efficient methods of identifying recombinant colonies after transformation eliminating time-consuming plasmid preparation. In the large-scale screen method, the partial colonies of E. coli bacteria can be directly transfered from the transformation plates into the solutions of the kits and immediately run on an agarose gel electrophoresis. The results will be known within 1-2 hours instead of the 2 days required in a conventional method of labor-intensive plasmid DNA miniprep or colony hybridization.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.