Learn More
Zymo Research Corporation DNA Clean & Concentrator-5(Capped), 50 Preps
Supplier: Zymo Research Corporation D4013
It can also be used for post-RT cDNA clean-up andpurification of sequencing-ready DNA from M13 phage. Simply add the specially formulated DNA Binding Buffer to your sample and transfer the mixture to the supplied Zymo-Spin™ Column. There is no need for organic denaturants or chloroform. Instead, the product features Fast-Spin column technology to yield DNA that is free of salts and contaminants in just 2 minutes. The purified DNA is ideal for DNA ligation, sequencing, labeling, PCR, microarray, transfection, transformation, and restriction digestion procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.