Learn More
Dojindo Molecular Technologies Inc DNA DAMAGE QUANTIFIC KIT (20 SAMPLES) is a colorimetric microplate assay and can determine the number of abasic sites in genomic DNA samples.
Supplier: Dojindo Molecular Technologies Inc DK0212