Learn More
Gold Biotechnology Inc dNTP Mix 1 mL
SDP
Supplier:  Gold Biotechnology Inc D9001
Encompass_Preferred
GoldBio’s 2’-Deoxynucleoside 5’-triphosphate mix is an aqueous solution containing dATP, dCTP, dGTP and dTTP, each at a final concentration of 10mM. This mixture has been tested for performance in a wide size-range of PCR templates with Taq and Pfu DNA polymerases.
Catalog No. NC2943340
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.