Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecyl Acetate 95.0+%, TCI America™
SDP

Catalog No. p-7117121 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 mL
500 mL

Chemical Identifiers

CAS 112-66-3
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.376
MDL Number MFCD00008973
InChI Key VZWGRQBCURJOMT-UHFFFAOYSA-N
Synonym lauryl acetate, acetic acid, dodecyl ester, dodecanol acetate, n-dodecyl acetate, 1-dodecanol acetate, dodecanyl acetate, acetate c-12, n-dodecyl ethanoate, dodecan-1-yl acetate, lauryl ethanoate
PubChem CID 8205
IUPAC Name dodecyl acetate
SMILES CCCCCCCCCCCCOC(=O)C

Specifications

Color Colorless
Boiling Point 180°C
Quantity 25 mL
Formula Weight 228.38
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dodecyl Acetate
EINECSNumber (2)-0735
RTECSNumber AH3525000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.