Learn More
TARGETMOL CHEMICALS INC DOFETILIDE 25MG
Supplier:  TARGETMOL CHEMICALS INC T6476
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dofetilide (UK 68789) is a sulfonamide class III antiarrhythmic agent and potassium channel blocker. Dofetilide selectively blocks cardiac ion channels of the rapid component of the delayed rectifier potassium current Ikr. This antiarrhythmic agent prolongs cardiac action potential duration and effective refractory period due to delayed repolarization without affecting conduction velocity. This results in a normal sinus rhythm. Dofetilide is used in the treatment of atrial fibrillation and flutter. purity: 99%
Catalog No. 50-257-6509
missing translation for 'QuantityLeft'