Learn More
MilliporeSigma™ Supelco™ domnick hunter™ Zero Air Generator
Produces a continuous supply of air that is virtually free of hydrocarbons, including methane
Supplier:  MilliporeSigma™ Supelco™ 28358U
 View more versions of this product
Catalog No. 11-100-5813
Add to Cart
Description
Description
  • Employs a catalytic oxidation system to produce a continuous supply of air that is virtually free of hydrocarbons, including methane
  • Incoming compressed air is filtered to remove oil, water, and particles
  • Filtered air is passed over a heated platinum catalyst that converts methane and oxygen in the air to carbon dioxide and water
  • If the hydrocarbon content of the input air is less than 10 ppm (as methane), the residual hydrocarbon concentration (as methane) is less than 0.1 ppm
  • A high efficiency 0.01 μm filter downstream of the catalyst ensures further removal of particles from the product stream
  • Maintenance filter kits and warranty information available upon request
  • Low power usage
  • Easy to wall mount
  • CE approved
Specifications
Specifications
Air Generator
CE Approved
230 V
10
Producing a Continuous Supply of Air
1000 cc/min
0.01 µm Filter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Title
Select an issue
domnick hunter is a registered trademark of Parker Hannifin