Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dopamine-1,1,2,2-d4 hydrochloride, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013467 Shop All MilliporeSigma Supelco Products
Click to view available options
:
5 mg

Specifications

Quantity 5 mg
CAS 203633-19-6
Type Analytical Standard
Linear Formula C8D4H7O2 N.HCl
MDL Number MFCD01317352
Synonym 2-(3,4-Dihydroxyphenyl)ethyl-1,1,2,2-d4-amine hydrochloride
Formula Weight 193.66
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Melting Point 248°C to 250°C
Physical Form Neat
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.