Learn More
Lkt Laboratories CAS 62-31-7, ≥99%, Endogenous dopamine D1-5 receptor agonist involved in motor control, mood, cognition, reward, and nausea. It also activates naïve and resting T cells and inhibits activated T cells.
SDP
Supplier:  Lkt Laboratories D5662
Encompass_Preferred
Dopamine is an endogenous hormone neurotransmitter that binds and activates dopamine receptors. Dopamine binding to D1/2/5 receptors increases levels of cAMP and dopamine binding to D3/4 receptors decreases levels of cAMP. Dopamine plays a role in motor control, mood, cognition, reward signaling, nausea, and arousal. Dopamine also exhibits immunomodulatory activity, activating naïve and resting T cells and inhibiting activated T cells.
Catalog No. NC2522163
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.