Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dopamine Receptor D4 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$173.10 - $403.50
Specifications
Antigen | Dopamine Receptor D4 |
---|---|
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Dopamine Receptor D4 Polyclonal antibody specifically detects Dopamine Receptor D4 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Dopamine Receptor D4 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
GPCR, Neuroscience | |
PBS with 50% glycerol, pH7.3. | |
1815 | |
IgG | |
Affinity purified |
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
Polyclonal | |
Purified | |
RUO | |
Mouse, Rat | |
D(2C) dopamine receptor, D(4) dopamine receptor, D4DR, Dopamine D4 receptor, dopamine receptor D4, seven transmembrane helix receptor | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human DRD4 (NP_000788.2). AWLLSPRLCDALMAMDVMLCTASIFNLCAISVDRFVAVAVPLRYNRQGGSRRQLLLIGATWLLSAAVAAPVLCGLNDVRGRDPAVCRLEDRDYVVYSSVCS | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title