Learn More
Medchemexpress LLC HY-B0109A 10mg Medchemexpress, Dorzolamide (hydrochloride) CAS:130693-82-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0109A10MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-B0109A 10mg Dorzolamide (hydrochloride) CAS:130693-82-2 Dorzolamide (L671152) hydrochloride is a potent carbonic anhydrase II inhibitor, with IC50 values of 0.18 nM and 600 nM for red blood cell CA-II and CA-I respectively. Dorzolamide possesses anti-tumor activity.
Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2365
missing translation for 'QuantityLeft'