Learn More
3M™ Double Coated Urethane Foam Tape
Shop All 3M Products- High shear adhesive with high temperature resistance
- Durable foam is ideal for interior mounting applications
- Ideal for bonding, attaching, and mounting
- Thick foam dampens and prevents vibration and open-cell structure aids component cooling
Specifications
For Use With (Application)
Commercial Solutions, Design and Construction, Manufacturing, Transportation
Adhesive Type
Acrylic
Color
Off-white
Description
Flexible
Dimensions (L x W)
36 yd. x 1 in.
Material
Urethane Foam
Model
4016
Quantity
9 Rolls/Cs.
Thickness (English)
62 mil
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.