Learn More
Foxx Life Sciences Borosil® Double Surface Condenser with Interchangeable Ground Joint 19/26, Jacket 300 mm, 5/CS
Supplier: Foxx Life Sciences 2641090
Borosil® Double Surface Condensers have a water jacket on the outside and through the center of the condensing tube. This is used when the liquid is very unstable like alcohol, aromatics, essential oils, petroleum products, distilling ethers, etc. These condensers are widely used throughout research laboratories. Avail. in Jacket size of 200-300 mm. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Low-Expansion Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.