Learn More
Selleck Chemical LLC Doxifluridine 25mg 3094-09-5 5'-DFUR, AMC 0101
Supplier:  Selleck Chemical LLC S204525MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Doxifluridine (5'-DFUR, AMC 0101) is an oral prodrug that is converted to the cytotoxic agent 5-fluorouracil (5-FU).
Catalog No. 50-136-1934