Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Doxorubicin hydrochloride, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Doxorubicin hydrochloride is a 14-hydroxylated version of Daunorubicin (Cat. No. 1467) that is naturally fluorescent. Doxorubicin is an antitumor antibiotic agent that inhibits DNA topoisomerase II. It is a DNA intercalator that inhibits nucleic acid synthesis and induces apoptosis. Doxorubicin reduces intracellular tau levels. Doxorubicin also promotes formation of free radicals for the disruption of membrane lipids and DNA strands. Doxorubicin fluorescence is quenched upon intercalation into the DNA; while binding to histones or partitioning into the phospholipid phase of PEG-phospholipid micelles or hydrophobic cores of polymeric micelles, increases Doxorubicin fluorescence. Doxorubicin fluorescence enables the monitoring of the localization of the drug within lipid bilayers and liposomal delivery systems and interaction of the drug with DNA and other macromolecules, as well as drug efflux pump activities.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 579.99 |
Percent Purity | >98% |
Chemical Name or Material | Doxorubicin hydrochloride |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction