Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Radians DPG62 Interceptor™ Compact Folding Earmuff
SDP

The DEWALT Interceptor Earmuff is a lightweight earmuff with unmatched performance providing maximum hearing protection

Supplier:  Radians DPG62C

Catalog No. 19-003-080



missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart

Description

Description

  • Adjustable moisture wicking headband moves moisture away while providing cool comfort
  • Lightweight design provides all day comfort
  • Compact folding feature allows for easy storage and keeps dust and debris out of the earcups
  • Cushion padding on earcups provides maximum protection from noise and maximum comfort
Specifications

Specifications

Foam
10.5 x 9.5 x 4.5 in.
Job Site Applications Including Shooting, Hunting, Construction, Manufacturing, Oil and Gas, Warehousing, Lawn and Garden, Automotive, Assembly, Public Safety, Home and DIY, or Utilities
1 Pk.
29 dB
Reusable
Black
Yes
Black
Passive Ear Muff
Interceptor™
0.6938 lb.
No
No
No
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

WARNING: Cancer and reproductive harm - www.P65warnings.ca.gov