Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DPN, Tocris Bioscience™
GSA_VA

Catalog No. p-4783434 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Highly potent ERβ agonist

Highly potent estrogen ERβ receptor agonist with a 70-fold selectivity over ERα (EC50 values are 0.85 and 66 nM respectively). Relaxes mesenteric arteries in vitro. Shown to regulate expression of GluR1, GluR2 and GluR3 in rat hippocampus.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 1428-67-7
Molecular Formula C15H13NO2
Molecular Weight (g/mol) 239.274
InChI Key GHZHWDWADLAOIQ-UHFFFAOYSA-N
Synonym diarylpropionitrile, 2,3-bis 4-hydroxyphenyl propionitrile, 2,3-bis 4-hydroxyphenyl propanenitrile, 2,3-bis 4-hydroxyphenyl-propionitrile, 2,3-bis p-hydroxyphenyl propionitrile, dpn compound, benzenepropanenitrile,4-hydroxy-a-4-hydroxyphenyl, diarylpropionitril, 2,3-bis p-hydroxyphenyl-propionitrile, propionitrile, 2,3-bis p-hydroxyphenyl
PubChem CID 102614
ChEBI CHEBI:63949
IUPAC Name 2,3-bis(4-hydroxyphenyl)propanenitrile
SMILES C1=CC(=CC=C1CC(C#N)C2=CC=C(C=C2)O)O

Specifications

Quantity 10 mg
Formula Weight 239.27
Percent Purity >99%
Chemical Name or Material DPN
Recommended Storage Desiccate at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.