Learn More
TARGETMOL CHEMICALS INC DPPH 200MG
Supplier:  TARGETMOL CHEMICALS INC T11093
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DPPH (22-Diphenyl-1-picrylhydrazyl) is a cell-permeable stable free radical that is commonly used to evaluate the ability of compounds to act as free radical scavengers or hydrogen donors and to measure the antioxidant activity of tissue extractspurity: 99%
Catalog No. 50-258-1577
missing translation for 'QuantityLeft'