Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DQP 1105, Tocris Bioscience™

Catalog No. p-4816609 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

GluN2C/GluN2D-selective NMDA antagonist

Noncompetitive NMDA receptor antagonist; displays over 50-fold selectivity for GluN2D- and GluN2C-containing receptors over GluN2B-, GluK2-, GluA1- and GluN2A-containing receptors (IC50 values are 2.7, 8.5, 121, 153, 198 and 206 μM, respectively). Reduces frequency of channel opening.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Chemical Identifiers

CAS 380560-89-4
Molecular Formula C29H24BrN3O4
Molecular Weight (g/mol) 558.432
InChI Key CADIBCWPGCNUBD-WEMUOSSPSA-N
Synonym 4-5-4-bromophenyl-3-2-hydroxy-6-methyl-4-phenylquinolin-3-yl-4,5-dihydro-1h-pyrazol-1-yl-4-oxobutanoic acid, 4-5-4-bromophenyl-3-6-methyl-2-oxo-4-phenyl-1,2-dihydroquinolin-3-yl-4,5-dihydro-1h-pyrazol-1-yl-4-oxobutanoic acid, 5-4-bromophenyl-3-1,2-dihydro-6-methyl-2-oxo-4-phenyl-3-quinolinyl-4,5-dihydro-gamma-oxo-1h-pyrazole-1-butanoic acid
PubChem CID 44657732
IUPAC Name 4-[(3E)-5-(4-bromophenyl)-3-(6-methyl-2-oxo-4-phenylquinolin-3-ylidene)pyrazolidin-1-yl]-4-oxobutanoic acid
SMILES CC1=CC2=C(C(=C3CC(N(N3)C(=O)CCC(=O)O)C4=CC=C(C=C4)Br)C(=O)N=C2C=C1)C5=CC=CC=C5

Specifications

Quantity 10 mg
Formula Weight 558.42
Percent Purity >97%
Chemical Name or Material DQP 1105
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.