Learn More
Eurofins Discoverx Ready-to-Assay D1 Dopamine Receptor Frozen Cells, 2 Vials of Cells. (HTS102RTA)
Supplier: Eurofins Discoverx HTS102RTA
The Ready-to-Assay D1 Dopamine Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. dopamine) based activation of DRD1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses DRD1 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of DRD1, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.