Learn More
Eurofins Discoverx ChemiScreen D4 Dopamine Receptor Stable Cell Line, 2 vials. (HTS223C)
Supplier: Eurofins Discoverx HTS223C
The ChemiScreen D4 Dopamine Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. dopamine) based activation of DRD4 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses DRD4 alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of DRD4, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.