Learn More
Sklar Instruments Forceps, Potts-Smith TC Dressing Forceps, Sklar 8-1/4 in., Straight, Cross-Serrated, Thumb, Tungsten Carbide / Stainless Steel, EA.
SDP
Supplier:  Sklar Instruments 514382
Encompass_Preferred
Forceps, Potts-Smith TC Dressing Forceps, Sklar, Straight, Cross-Serrated, Thumb, Tungsten Carbide / Stainless Steel, EA. Sklar Potts-Smith Tungsten Carbide Dressing Forceps are most commonly used for application and changing of dressings. They may also be used for dissection of fine or single-layer tissue. Cross-serrated tips provide a solid grip on both materials and tissue. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles. This product is straight with cross-serrated tips and a length of 8-1/4 inches. Lifetime warranty against all manufacturer defects.
Catalog No. 50-110-9618
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.