Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Calibrators

Catalog No. p-6225194 Shop All Thermo Scientific Products
Click to view available options
Concentration:
This item is not returnable. View return policy
This item is not returnable. View return policy

Save time while maintaining accuracy with the liquid, ready-to-use Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Calibrators.

Ensure accuracy and precision of your drug testing results by partnering these liquid, ready-to-use Thermo Scientific™ DRI™ Hydrocodone Drugs of Abuse Calibrators with our Thermo Scientific DRI Hydrocodone Drugs of Abuse Assay and Control Set.These calibrators are for sale in the U.S.A. and Canada only.

  • Specific detection of hydrocodone
  • Qualitative or semi-quantitative modes
  • For sale in the U.S.A. and Canada only



Specifications

Content And Storage

2°C to 8°C

Description

DRI Hydrocodone Drugs of Abuse Calibrator

Concentration

100 ng/mL

Detectable Analytes

Hydrocodone

Sample Type

Urine

Type

Calibrator

Packaging

1 Bottle

Quantity

1 x 10 mL

Disclaimers:
Some of these products are not cleared in every country. Please contact your sales representative for more information. Product claims may differ from country to country based on regulations and product approvals. Please contact your country representative for further details. 

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.