Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Methaqualone Drugs of Abuse Assays

Save time in reagent preparation with Thermo Scientific™ DRI™ Methaqualone Drugs of Abuse Assays.

$2620.00

Specifications

Control Sets MGC Multi-Drug Controls
Description DRI Methaqualone Drugs of Abuse Assays
DoA Calibrators DRI Multi-Drug Calibrators
Content And Storage 2°C to 8°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
23-278-324
View Documents
Thermo Scientific™
0514
1 x 100 mL
Each of 1 for $2,620.00
Add to Cart
 
Description

Description

Save time in reagent preparation with Thermo Scientific™ DRI™ Methaqualone Drugs of Abuse Assays, which offer applications for an array of clinical chemistry analyzers. These liquid, ready-to-use assays have been field-tested on a wide range of analyzers toeliver high analytical accuracy. They are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.

  • Qualitative or semi-quantitative options for results
  • Excellent correlation to GC/MS
  • Variety of calibrators and multiconstituent control sets
Specifications

Specifications

MGC Multi-Drug Controls
DRI Methaqualone Drugs of Abuse Assays
DRI Multi-Drug Calibrators
2°C to 8°C
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.