Learn More
Cole-Parmer Drierite 23001 Indicating desiccant; twelve 1-lb packs/carton
Supplier:  Cole-Parmer 0719305
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Use Drierite desiccants -- anhydrous calcium sulfate in desiccators, drying columns or drying tubes. Choose indicating or non-indicating desiccant indicating desiccant starts blue, then turns pink when regeneration or renewal is necessary.
Catalog No. NC1501597