Learn More
Stoelting™ Drill and Injection Robot for Motorized Stereotaxic Instruments
Provides for a fully software driven solution to stereotaxic surgery
Supplier: Stoelting™ 58645
Description
- Allows for high precision injection
- After loading the syringe, the injection robot allows for multiple injections in different animals without tool exchange or recalibration
- Integrated into the StereoDrive software (computer and motorized stereotaxic not included but required)
- Available as Glass-Capillary Nanoinjector
- Atlas Integration
- High Throughput Drilling and Injection
- No Tool Exchange
- Defined/Stored Protocols
- Multisite Microinjections
- Ideal for Optogenetics applications
- Precise Depth Control
- Brain Windowing
- Contour Drilling
- Smooth Edges
- Skull Thinning
Specifications
Neuroscience | |
Injection Robot for Motorized Stereotaxic Systems |