Learn More
Quantimetrix Urinalysis Control, Point-of-care. Liquid, made with human urine, has a 1-month open vial stability at room temperature. 18-month shelf life & open vial stability from date of manufacture when stored at 2-8 deg. C. 2 level set, 2x5mL. 57-55-6
SDP
Supplier:  Quantimetrix 144006
Encompass_Preferred
Urinalysis Control, Point-of-care. Liquid, made with human urine, has a 1-month open vial stability at room temperature. 18-month shelf life & open vial stability from date of manufacture when stored at 2-8 deg. C. 2 level set, 2x5mL. 57-55-6
Catalog No. 50-152-8627
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.